Learn More
Thomas Scientific 96 Round Well Microplate, Low Binding; 200 æl round wells, V-shaped bottom, clear PP, 50 plates
SDP
Catalog No. NC1640990
Encompass
These shallow 96 well storage microplates are particularly suitable for collection and preservation of samples widely used in cell biology research, molecular biology research, and drug discovery. Four formats are available of this plate - a U-bottom plate with 300 µl or 330 µl well volumes (4ti-0110 and 4ti-0116 respectively), and a V-bottom plate with 200 µl or 330 µl well volume (4ti-LB0109 and 4ti-0117 respectively). Now also available: a Low Binding plate with ultra-low DNA binding properties (4ti-LB0109) for sensitive applications such as Next Generation Sequencing sample preparation. All of these plates are ultra-flat for a completely uniform seal; either with adhesive or heat seals, or with our accompanying silicone sealing mat (4ti-0138). The plates are manufactured in cleanroom facilities which are certified free of RNase, DNase, human genomic DNA and endotoxins. We use only the highest medical grade virgin polypropylene with high resistance against ch
Catalog No. NC1640990 Supplier Thomas Scientific Supplier No. 1149Q98
These shallow 96 well storage microplates are particularly suitable for collection and preservation of samples widely used in cell biology research, molecular biology research, and drug discovery. Four formats are available of this plate - a U-bottom plate with 300 µl or 330 µl well volumes (4ti-0110 and 4ti-0116 respectively), and a V-bottom plate with 200 µl or 330 µl well volume (4ti-LB0109 and 4ti-0117 respectively). Now also available: a Low Binding plate with ultra-low DNA binding properties (4ti-LB0109) for sensitive applications such as Next Generation Sequencing sample preparation. All of these plates are ultra-flat for a completely uniform seal; either with adhesive or heat seals, or with our accompanying silicone sealing mat (4ti-0138). The plates are manufactured in cleanroom facilities which are certified free of RNase, DNase, human genomic DNA and endotoxins. We use only the highest medical grade virgin polypropylene with high resistance against ch