Learn More
Cellvis 96WELL PLT W/0 CVER GLS IND 20
Catalog No. NC1034695
Encompass
96 well Glass Bottom Plate with #0 cover glass
Catalog No. NC1034695 Supplier Cellvis Supplier No. P96100N
96 well Glass Bottom Plate with #0 cover glass