Learn More
Thomas Scientific 96W Cellstar Plate, PS, TC Treated, Sterile, Flat Bottom, Chimney Style, Black uClear, w/ Lid, 32/cs
SDP
Catalog No. 50320785
Encompass_Preferred
CELLSTAR format sterile plates are ideal for cell culture applications and are available tissue culture treated for the culture of anchorage-dependent cells, or untreated for suspension culture applications.
This item is not returnable. View return policy
Catalog No. 50-320-785 Supplier Thomas Scientific Supplier No. 1184J28
CELLSTAR format sterile plates are ideal for cell culture applications and are available tissue culture treated for the culture of anchorage-dependent cells, or untreated for suspension culture applications.