Learn More
CELLTREAT Scientific Products 1.5mL 96 Deep Well Storage Plate, PP, Square Well, Round Bottom, Non-sterile
SDP
Catalog No. 501149076
Encompass_Preferred
CELLTREAT Scientific Products 96 Deep Well Storage Plate. 96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications. Square to round transition of V-Bottom maximizes working volume. Well volume of 1.5mL, square well shape with a round bottom. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/bag, 25/case.
Catalog No. 50-114-9076 Supplier CELLTREAT Scientific Products Supplier No. 229573
CELLTREAT Scientific Products 96 Deep Well Storage Plate. 96 Deep Well Storage Plates can be used for the growth of bacterial cultures or storage of compounds. Polypropylene offers a low binding surface to prevent samples from sticking to the side walls during elution, and is chemically inert for combinatorial chemistry applications. Square to round transition of V-Bottom maximizes working volume. Well volume of 1.5mL, square well shape with a round bottom. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/bag, 25/case.