Learn More
Cayman Chemical 9AmIno6chloro2methoxyacr 10mg
SDP
Catalog No. 502986847
Encompass_Preferred
A fluorescent probe for DNA and pH gradients; selective for poly(dA-dT) sequences; has ex/em spectra of 411/475 nm, respectively; inhibits AChE (Ki = 49 nM)
Catalog No. 50-298-6847 Supplier Cayman Chemical Supplier No. 2243910 MG
A fluorescent probe for DNA and pH gradients; selective for poly(dA-dT) sequences; has ex/em spectra of 411/475 nm, respectively; inhibits AChE (Ki = 49 nM)