Learn More
Cayman Chemical 9deoxy9methylen16 16dIm 10mg
SDP
Catalog No. 502971543
Encompass_Preferred
An impurity generated in the production of travoprost a 496 diastereomeric mixture of epoxides
Catalog No. 50-297-1543 Supplier Cayman Chemical Supplier No. 1443010 MG
An impurity generated in the production of travoprost a 496 diastereomeric mixture of epoxides