Learn More
Cayman Chemical 9deoxy9methylen16 16dIm 10mg
SDP
Catalog No. 502969358
Encompass_Preferred
9-deoxy-9-methylene-16,16-dimethyl Prostaglandin E2 (Meteneprost) is a potent analog of prostaglandin E2 (PGE2; Item No. 14010) with an extended half-life in vivo. In combination with various other prostaglandin derivatives, it results in the termination of first trimester pregnancy in monkeys. A single intramuscular injection containing 0.5 mg of meteneprost and 7.5 mg of 17-phenyl trinor PGF1α is very effective in terminating EArly pregnancy.{4692} This prostaglandin mixture is ineffective on monkeys in their third trimester of pregnancy.{4692} Meteneprost, when compared to PGE2 and PGF1α, in monkey and rat, does not result in unwanted side effects such as fever or gastrointestinal problems.{4692,464}
Catalog No. 50-296-9358 Supplier Cayman Chemical Supplier No. 1442010 MG
9-deoxy-9-methylene-16,16-dimethyl Prostaglandin E2 (Meteneprost) is a potent analog of prostaglandin E2 (PGE2; Item No. 14010) with an extended half-life in vivo. In combination with various other prostaglandin derivatives, it results in the termination of first trimester pregnancy in monkeys. A single intramuscular injection containing 0.5 mg of meteneprost and 7.5 mg of 17-phenyl trinor PGF1α is very effective in terminating EArly pregnancy.{4692} This prostaglandin mixture is ineffective on monkeys in their third trimester of pregnancy.{4692} Meteneprost, when compared to PGE2 and PGF1α, in monkey and rat, does not result in unwanted side effects such as fever or gastrointestinal problems.{4692,464}