Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9H-carbazol-4-ol | 52602-39-8 | MFCD02178385 | 99.3% | 183.21 g/mol | C12H9NO | 100 G
SDP

Catalog No. 5000302190
Encompass_Preferred

9H-Carbazol-4-ol (4-hydroxycarbazole) is an aromatic heterocyclic compound commonly used as a building block in organic synthesis and materials research. It is suitable for laboratory synthesis, analytical assays, and development of heterocyclic derivatives. Handle and store according to supplier safety data guidelines.

  • High purity (99.29%) appropriate for synthesis and analysis.
  • Chemical formula C12H9NO and molecular weight 183.21 g/mol.
  • CAS registry number 52602-39-8 for unambiguous identification.
  • Available in multiple laboratory pack sizes, including 100 g, for scale flexibility.
  • Synonym: 4-hydroxycarbazole, useful when searching literature and databases.

Catalog No. 50-003-02190 Supplier Medchemexpress LLC Supplier No. HYW016331100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9H-Carbazol-4-ol (4-hydroxycarbazole) is an aromatic heterocyclic compound commonly used as a building block in organic synthesis and materials research. It is suitable for laboratory synthesis, analytical assays, and development of heterocyclic derivatives. Handle and store according to supplier safety data guidelines.

  • High purity (99.29%) appropriate for synthesis and analysis.
  • Chemical formula C12H9NO and molecular weight 183.21 g/mol.
  • CAS registry number 52602-39-8 for unambiguous identification.
  • Available in multiple laboratory pack sizes, including 100 g, for scale flexibility.
  • Synonym: 4-hydroxycarbazole, useful when searching literature and databases.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.