Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9H-carbazol-4-ol | 52602-39-8 | MFCD02178385 | 99.3% | 183.21 g·mol⁻¹ | C12H9NO | 500 G
SDP

Catalog No. 5000302239
Encompass_Preferred

9H-carbazol-4-ol (4-hydroxycarbazole) is an aromatic heterocyclic compound used in organic synthesis and medicinal chemistry research. Supplied as a solid with high analytical purity and available in laboratory pack sizes, it is commonly used as a building block in heterocycle construction and functionalization studies.

  • High purity (99.3%).
  • Solid, off-white to light brown appearance.
  • Molecular formula C12H9NO; molecular weight 183.21 g·mol⁻¹.
  • Suitable for organic synthesis and heterocycle chemistry.
  • Available in multiple pack sizes, including 500 g.
  • Analytical documentation available (COA, SDS).

Catalog No. 50-003-02239 Supplier Medchemexpress LLC Supplier No. HYW016331500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9H-carbazol-4-ol (4-hydroxycarbazole) is an aromatic heterocyclic compound used in organic synthesis and medicinal chemistry research. Supplied as a solid with high analytical purity and available in laboratory pack sizes, it is commonly used as a building block in heterocycle construction and functionalization studies.

  • High purity (99.3%).
  • Solid, off-white to light brown appearance.
  • Molecular formula C12H9NO; molecular weight 183.21 g·mol⁻¹.
  • Suitable for organic synthesis and heterocycle chemistry.
  • Available in multiple pack sizes, including 500 g.
  • Analytical documentation available (COA, SDS).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.