Learn More
Medchemexpress LLC 9h-Carbazole-3-methanamine, 9-ethyl-N-methyl-, hydrochloride (1:1) | 1050480-30-2 | 99.3% | 274.79 | 1 ML
SDP
Catalog No. 5000344328
Encompass_Preferred
PhiKan 083 hydrochloride is a carbazole derivative that binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM, and a relative binding affinity (Kd) of 150 μM in Ln229 cells.
  • Slows down its thermal denaturation rate.
Catalog No. 50-003-44328 Supplier Medchemexpress LLC Supplier No. HY108637A10MG
PhiKan 083 hydrochloride is a carbazole derivative that binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM, and a relative binding affinity (Kd) of 150 μM in Ln229 cells.
  • Slows down its thermal denaturation rate.