Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9H-Carbazole-9-ethanol, 3,6-dibromo-α-[[(3-methoxyphenyl)amino]methyl]- | 313268-18-7 | 98.26% | 100 MG
SDP

Catalog No. 5000338978
Encompass_Preferred

P7C3-OMe is a pro-neurogenic compound that offers therapeutic benefits in neuropsychiatric and/or neurodegenerative diseases. The R-enantiomer of P7C3-OMe is significantly more active than its S-enantiomer.

  • Pro-neurogenic compound
  • Offers therapeutic benefits in neuropsychiatric and neurodegenerative diseases
  • R-enantiomer is significantly more active than S-enantiomer
  • Molecular formula: C22H20Br2N2O2
  • Molecular weight: 504.21
  • CAS number: 313268-18-7
  • Appearance: Solid, white to off-white
  • Purity: 98.26%
  • Soluble in DMSO at 100 mg/mL or greater

Catalog No. 50-003-38978 Supplier Medchemexpress LLC Supplier No. HY128856100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

P7C3-OMe is a pro-neurogenic compound that offers therapeutic benefits in neuropsychiatric and/or neurodegenerative diseases. The R-enantiomer of P7C3-OMe is significantly more active than its S-enantiomer.

  • Pro-neurogenic compound
  • Offers therapeutic benefits in neuropsychiatric and neurodegenerative diseases
  • R-enantiomer is significantly more active than S-enantiomer
  • Molecular formula: C22H20Br2N2O2
  • Molecular weight: 504.21
  • CAS number: 313268-18-7
  • Appearance: Solid, white to off-white
  • Purity: 98.26%
  • Soluble in DMSO at 100 mg/mL or greater

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.