Learn More
Medchemexpress LLC 9H-fluoren-2-amine | 153-78-6 | MFCD00001125 | 95.0% | 181.24 g/mol | C13H11N | 5 G
SDP
Catalog No. 5000301136
Encompass_Preferred
2-Aminofluorene is a research-grade arylamine genotoxin used in laboratory studies of DNA adduct structure, DNA repair, mutagenesis, and carcinogenesis. It is supplied as a solid for standard research applications.
  • Used for DNA adduct, DNA repair, mutagenesis, and carcinogenesis research.
  • Chemical formula C13H11N, molecular weight 181.24 g/mol.
  • 95.0% purity.
  • Available in multiple pack sizes including a 5 g option.
  • Solid form; follow SDS and COA for recommended storage and handling.
Catalog No. 50-003-01136 Supplier Medchemexpress LLC Supplier No. HYW0164335G
2-Aminofluorene is a research-grade arylamine genotoxin used in laboratory studies of DNA adduct structure, DNA repair, mutagenesis, and carcinogenesis. It is supplied as a solid for standard research applications.
  • Used for DNA adduct, DNA repair, mutagenesis, and carcinogenesis research.
  • Chemical formula C13H11N, molecular weight 181.24 g/mol.
  • 95.0% purity.
  • Available in multiple pack sizes including a 5 g option.
  • Solid form; follow SDS and COA for recommended storage and handling.