Learn More
Medchemexpress LLC 9H-Fluoren-2-amine | 153-78-6 | MFCD00001125 | 99.9% | 181.24 | C13H11N | 100 G
SDP
Catalog No. 5000302188
Encompass_Preferred
2-Aminofluorene is an aromatic amine research reagent and genotoxin used to study DNA adduct formation, DNA repair, carcinogenesis, and mutagenesis. It is provided as a solid in multiple pack sizes and as a solution in DMSO for biochemical assays.
  • Used to investigate DNA adduct structure and mutagenesis.
  • Useful in studies of DNA repair and carcinogenesis.
  • Available as solid material and as a 10 mM solution in DMSO.
  • High purity suitable for analytical and biological research.
  • Supplied with a product datasheet and certificate of analysis.
Catalog No. 50-003-02188 Supplier Medchemexpress LLC Supplier No. HYW016433100G
2-Aminofluorene is an aromatic amine research reagent and genotoxin used to study DNA adduct formation, DNA repair, carcinogenesis, and mutagenesis. It is provided as a solid in multiple pack sizes and as a solution in DMSO for biochemical assays.
  • Used to investigate DNA adduct structure and mutagenesis.
  • Useful in studies of DNA repair and carcinogenesis.
  • Available as solid material and as a 10 mM solution in DMSO.
  • High purity suitable for analytical and biological research.
  • Supplied with a product datasheet and certificate of analysis.