Learn More
Medchemexpress LLC 9H-fluoren-2-amine | 153-78-6 | MFCD00001125 | 99.9% | 181.24 g/mol | C13H11N | 25 G
SDP
Catalog No. 5000303796
Encompass_Preferred
2-Aminofluorene (CAS 153-78-6) is an aromatic amine used as a genotoxin probe in research on DNA adduct formation, DNA repair, carcinogenesis, and mutagenesis. It is supplied as an off-white to light-yellow solid with analytical-grade purity for biochemical and molecular biology applications; handle with appropriate safety precautions.
  • High purity (99.9%).
  • Off-white to light yellow solid for easy identification.
  • Stable as powder at -20°C for long-term storage.
  • Soluble in common organic solvents for stock solution preparation.
  • Available in multiple pack sizes to support small-scale and bulk experiments.
  • Suitable for studies of DNA adducts, DNA repair, carcinogenesis, and mutagenesis.
Catalog No. 50-003-03796 Supplier Medchemexpress LLC Supplier No. 25GHYW016433
2-Aminofluorene (CAS 153-78-6) is an aromatic amine used as a genotoxin probe in research on DNA adduct formation, DNA repair, carcinogenesis, and mutagenesis. It is supplied as an off-white to light-yellow solid with analytical-grade purity for biochemical and molecular biology applications; handle with appropriate safety precautions.
  • High purity (99.9%).
  • Off-white to light yellow solid for easy identification.
  • Stable as powder at -20°C for long-term storage.
  • Soluble in common organic solvents for stock solution preparation.
  • Available in multiple pack sizes to support small-scale and bulk experiments.
  • Suitable for studies of DNA adducts, DNA repair, carcinogenesis, and mutagenesis.