Learn More
Medchemexpress LLC 9H-fluoren-2-ol-d9 (NSC 31248-d9) | 922510-18-7 | MFCD28899138 | 191.27 | C13HD9O | 1 MG
SDP
Catalog No. 5000302298
Encompass_Preferred
9H-Fluoren-2-ol-d9 is the deuterium-labeled form of 9H-fluoren-2-ol (CAS 922510-18-7), provided as a stable-isotope reference standard for analytical and drug-development workflows. The labeled compound (C13HD9O, MW 191.27) is intended for use as a tracer in quantitation, metabolite identification, and pharmacokinetic studies using mass spectrometry and related techniques.
  • Deuterium-labeled for use as a stable-isotope tracer.
  • Suitable for LC-MS quantitation and metabolite studies.
  • Molecular formula C13HD9O and molecular weight 191.27 aid method development.
  • Supplied at research-scale quantities appropriate for analytical standards.
  • Certificate of analysis and QC reports available on request.
Catalog No. 50-003-02298 Supplier Medchemexpress LLC Supplier No. HYW016389S1MG
9H-Fluoren-2-ol-d9 is the deuterium-labeled form of 9H-fluoren-2-ol (CAS 922510-18-7), provided as a stable-isotope reference standard for analytical and drug-development workflows. The labeled compound (C13HD9O, MW 191.27) is intended for use as a tracer in quantitation, metabolite identification, and pharmacokinetic studies using mass spectrometry and related techniques.
  • Deuterium-labeled for use as a stable-isotope tracer.
  • Suitable for LC-MS quantitation and metabolite studies.
  • Molecular formula C13HD9O and molecular weight 191.27 aid method development.
  • Supplied at research-scale quantities appropriate for analytical standards.
  • Certificate of analysis and QC reports available on request.