Learn More
Medchemexpress LLC 9H-fluoren-9-one-d8 | 137219-34-2 | 98.9% | 10 MG
SDP
Catalog No. 5000297596
Encompass_Preferred
9H-Fluoren-9-one-d8 is the deuterium-labeled analogue of fluorenone supplied as a solid isotopically labeled standard for analytical workflows. It functions as a tracer and internal standard to support accurate quantitation and method validation in research, environmental, and drug-development applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High isotopic enrichment with reported purity of 98.87%.
  • Solid form for straightforward handling and storage.
  • Available in small-mass pack sizes for analytical use.
Catalog No. 50-002-97596 Supplier Medchemexpress LLC Supplier No. HY32181S10MG
9H-Fluoren-9-one-d8 is the deuterium-labeled analogue of fluorenone supplied as a solid isotopically labeled standard for analytical workflows. It functions as a tracer and internal standard to support accurate quantitation and method validation in research, environmental, and drug-development applications.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High isotopic enrichment with reported purity of 98.87%.
  • Solid form for straightforward handling and storage.
  • Available in small-mass pack sizes for analytical use.