Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (((9H-fluoren-9-yl)methoxy)carbonyl)-D-histidine | 157355-79-8 | MFCD00237022 | 377.39 g/mol | C21H19N3O4 | 5 G
SDP

Catalog No. 5000209774
Encompass_Preferred

(((9H-fluoren-9-yl)methoxy)carbonyl)-D-histidine is an Fmoc-protected D-histidine derivative used as a protected amino acid building block for solid-phase peptide synthesis and peptide chemistry research.

  • Used as an Fmoc-protected D-histidine building block for peptide synthesis.
  • Molecular formula C21H19N3O4, molecular weight 377.39 g/mol.
  • CAS number 157355-79-8.
  • Available in multiple pack sizes up to 50 g.
  • Store at 4°C and protect from light; in solution, -80°C for up to 6 months, -20°C for up to 1 month.

Catalog No. 50-002-09774 Supplier Medchemexpress LLC Supplier No. HYW0487015G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(((9H-fluoren-9-yl)methoxy)carbonyl)-D-histidine is an Fmoc-protected D-histidine derivative used as a protected amino acid building block for solid-phase peptide synthesis and peptide chemistry research.

  • Used as an Fmoc-protected D-histidine building block for peptide synthesis.
  • Molecular formula C21H19N3O4, molecular weight 377.39 g/mol.
  • CAS number 157355-79-8.
  • Available in multiple pack sizes up to 50 g.
  • Store at 4°C and protect from light; in solution, -80°C for up to 6 months, -20°C for up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.