Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine | 110990-08-4 | MFCD04113545 | 99.5% | 368.43 g/mol | C21H24N2O4 | 1 G
SDP

Catalog No. 5000208629
Encompass_Preferred

(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is an Fmoc-protected D-lysine amino acid derivative used primarily as a building block in solid-phase peptide synthesis and related peptide chemistry applications. It is provided as a high-purity, crystalline compound suitable for coupling reactions and standard peptide synthesis workflows.

  • Fmoc-protected D-lysine suitable for peptide synthesis.
  • High-purity material for reliable coupling and characterization.
  • Formula: C21H24N2O4 and molecular weight 368.43 g/mol.
  • CAS 110990-08-4 for unambiguous identification.
  • Available in small laboratory quantities for research use.
  • Store protected from light at refrigerated temperatures; follow solvent storage guidance for long-term stability.

Catalog No. 50-002-08629 Supplier Medchemexpress LLC Supplier No. HYW0906261G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is an Fmoc-protected D-lysine amino acid derivative used primarily as a building block in solid-phase peptide synthesis and related peptide chemistry applications. It is provided as a high-purity, crystalline compound suitable for coupling reactions and standard peptide synthesis workflows.

  • Fmoc-protected D-lysine suitable for peptide synthesis.
  • High-purity material for reliable coupling and characterization.
  • Formula: C21H24N2O4 and molecular weight 368.43 g/mol.
  • CAS 110990-08-4 for unambiguous identification.
  • Available in small laboratory quantities for research use.
  • Store protected from light at refrigerated temperatures; follow solvent storage guidance for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.