Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine | 110990-08-4 | MFCD04113545 | 99.5% | 368.43 g/mol | C21H24N2O4 | 25 G
SDP

Catalog No. 5000209593
Encompass_Preferred

(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is an Fmoc-protected D-enantiomer of lysine used as a building block in solid-phase peptide synthesis and peptide chemistry research.

  • Fmoc-protected D-lysine for peptide synthesis.
  • Molecular weight 368.43 g/mol.
  • Chemical formula C21H24N2O4.
  • High reported purity (99.51%).
  • Recommended storage at 4°C; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
  • Off-white solid available in bulk pack sizes suitable for research use.

Catalog No. 50-002-09593 Supplier Medchemexpress LLC Supplier No. 25GHYW090626
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(((9H-fluoren-9-yl)methoxy)carbonyl)-D-lysine is an Fmoc-protected D-enantiomer of lysine used as a building block in solid-phase peptide synthesis and peptide chemistry research.

  • Fmoc-protected D-lysine for peptide synthesis.
  • Molecular weight 368.43 g/mol.
  • Chemical formula C21H24N2O4.
  • High reported purity (99.51%).
  • Recommended storage at 4°C; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
  • Off-white solid available in bulk pack sizes suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.