Learn More
Medchemexpress LLC (((9H-Fluoren-9-yl)methoxy)carbonyl)-D-tyrosine | 112883-29-1 | MFCD00171384 | 99.5% | 403.43 g/mol | C24H21NO5 | 25 G
SDP
Catalog No. 5000209500
Encompass_Preferred
(((9H-Fluoren-9-yl)methoxy)carbonyl)-D-tyrosine is an Fmoc-protected D-tyrosine used as a protected amino acid building block in solid-phase peptide synthesis. It is a white to off-white solid with high purity and good solubility in DMSO.
  • Used as a building block for solid-phase peptide synthesis.
  • Provides Fmoc protection for the amino terminus during coupling steps.
  • High reported purity suitable for research applications.
  • Soluble in DMSO at high concentration with ultrasonic assistance.
  • Stable when stored at recommended cold temperatures in powder form.
Catalog No. 50-002-09500 Supplier Medchemexpress LLC Supplier No. 25GHYW048703
(((9H-Fluoren-9-yl)methoxy)carbonyl)-D-tyrosine is an Fmoc-protected D-tyrosine used as a protected amino acid building block in solid-phase peptide synthesis. It is a white to off-white solid with high purity and good solubility in DMSO.
  • Used as a building block for solid-phase peptide synthesis.
  • Provides Fmoc protection for the amino terminus during coupling steps.
  • High reported purity suitable for research applications.
  • Soluble in DMSO at high concentration with ultrasonic assistance.
  • Stable when stored at recommended cold temperatures in powder form.