Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (((9H-Fluoren-9-yl)methoxy)carbonyl)-L-histidine | 116611-64-4 | MFCD00190885 | 99.7% | 377.39 g/mol | C21H19N3O4 | 25 G
SDP

Catalog No. 5000209503
Encompass_Preferred

(((9H-Fluoren-9-yl)methoxy)carbonyl)-L-histidine is an Fmoc-protected derivative of the amino acid L-histidine supplied as a solid for research use, commonly used as a building block in peptide synthesis and related chemical research.

  • Fmoc-protected amino acid suitable for peptide synthesis.
  • High purity: 99.7% (as supplied).
  • White to off-white solid appearance.
  • Molecular formula C21H19N3O4; molecular weight 377.39 g/mol.
  • Storage: powder stable at -20°C for up to 3 years, 4°C for up to 2 years; in solvent store at -80°C for up to 6 months.

Catalog No. 50-002-09503 Supplier Medchemexpress LLC Supplier No. 25GHYW048911
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(((9H-Fluoren-9-yl)methoxy)carbonyl)-L-histidine is an Fmoc-protected derivative of the amino acid L-histidine supplied as a solid for research use, commonly used as a building block in peptide synthesis and related chemical research.

  • Fmoc-protected amino acid suitable for peptide synthesis.
  • High purity: 99.7% (as supplied).
  • White to off-white solid appearance.
  • Molecular formula C21H19N3O4; molecular weight 377.39 g/mol.
  • Storage: powder stable at -20°C for up to 3 years, 4°C for up to 2 years; in solvent store at -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.