Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical (9H-fluoren-9-yl)methyl (2-(2-aminoethoxy)ethyl)carbamate hydrochloride | 10 g | CAS 221352-88-1 | MDL MFCD01321025

Catalog No. 502360086
Encompass

(9H-fluoren-9-yl)methyl (2-(2-aminoethoxy)ethyl)carbamate hydrochloride is a Amino Acid reagent (Subcategory: Building Block) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 221352-88-1
- MDL: MFCD01321025
- InChIKey: ZRXOWZYREIYUHG-UHFFFAOYSA-N
- Molecular Weight: 362.854
- Molecular Formula: C19H23ClN2O3
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl (2-(2-aminoethoxy)ethyl)carbamate hydrochloride
- SMILES: NCCOCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O.Cl

Catalog No. 50-236-0086 Supplier STA Pharmaceutical Supplier No. STAAA1092610GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(9H-fluoren-9-yl)methyl (2-(2-aminoethoxy)ethyl)carbamate hydrochloride is a Amino Acid reagent (Subcategory: Building Block) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 221352-88-1
- MDL: MFCD01321025
- InChIKey: ZRXOWZYREIYUHG-UHFFFAOYSA-N
- Molecular Weight: 362.854
- Molecular Formula: C19H23ClN2O3
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl (2-(2-aminoethoxy)ethyl)carbamate hydrochloride
- SMILES: NCCOCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O.Cl

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.