Learn More
Medchemexpress LLC 9H-Fluorene-9-methanol | 24324-17-2 | 99.0% | 196.25 | 100 G
SDP
Catalog No. 5000366390
Encompass_Preferred
9-Fluorenylmethanol is an N-protecting reagent primarily used in peptide synthesis. It serves a crucial role in safeguarding the amino group during various chemical reactions involved in constructing peptide chains. This compound ensures the integrity of the amino acid building blocks, allowing for controlled and efficient synthesis of complex peptides. Its application is significant in research settings where precise peptide structures are essential for studying biological functions and developing new therapeutic agents.
  • Used as an N-protecting reagent in peptide synthesis.
  • Helps protect the amino group during peptide formation.
  • Enables controlled synthesis of complex peptides.
  • Supports research requiring precise peptide structures.
Catalog No. 50-003-66390 Supplier Medchemexpress LLC Supplier No. HYW009876100G
9-Fluorenylmethanol is an N-protecting reagent primarily used in peptide synthesis. It serves a crucial role in safeguarding the amino group during various chemical reactions involved in constructing peptide chains. This compound ensures the integrity of the amino acid building blocks, allowing for controlled and efficient synthesis of complex peptides. Its application is significant in research settings where precise peptide structures are essential for studying biological functions and developing new therapeutic agents.
  • Used as an N-protecting reagent in peptide synthesis.
  • Helps protect the amino group during peptide formation.
  • Enables controlled synthesis of complex peptides.
  • Supports research requiring precise peptide structures.