Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9H-purin-6-amine, 9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl) | 20227-41-2 | MFCD12911815 | 99.9% | 269.24 g·mol⁻¹ | C10H12FN5O3 | 25 MG
SDP

Catalog No. 5000281559
Encompass_Preferred

2'-Deoxy-2'-fluoroarabinoadenosine is a fluorinated deoxyadenosine nucleoside analogue used as a research reagent. It has reported antitumor and antiviral activity and is supplied as a high-purity solid for laboratory studies.

  • Fluorinated deoxyadenosine nucleoside analogue.
  • Reported antitumor and antiviral activity.
  • Chemical formula C10H12FN5O3.
  • Molecular weight 269.24 g·mol⁻¹.
  • High purity (≈99.9% by LCMS).
  • Storage: 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-81559 Supplier Medchemexpress LLC Supplier No. HYW25168725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Deoxy-2'-fluoroarabinoadenosine is a fluorinated deoxyadenosine nucleoside analogue used as a research reagent. It has reported antitumor and antiviral activity and is supplied as a high-purity solid for laboratory studies.

  • Fluorinated deoxyadenosine nucleoside analogue.
  • Reported antitumor and antiviral activity.
  • Chemical formula C10H12FN5O3.
  • Molecular weight 269.24 g·mol⁻¹.
  • High purity (≈99.9% by LCMS).
  • Storage: 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.