Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9H-Purine, 2,6-dichloro-9-β-D-ribofuranosyl- | 13276-52-3 | 321.12 | 250 MG
SDP

Catalog No. 5000449883
Encompass_Preferred

2,6-Dichloropurine Riboside (CAS 13276-52-3) is a chemical compound with a molecular weight of 321.12 and a purity of 98.97%. It appears as a white to off-white solid and is intended for research use only.

  • Chemical name: 9H-Purine, 2,6-dichloro-9-β-D-ribofuranosyl-
  • CAS number: 13276-52-3
  • Molecular formula: C10H10Cl2N4O4
  • Molecular weight: 321.12
  • Purity: 98.97%
  • Appearance: White to off-white solid
  • Storage conditions: -20°C, sealed storage, away from moisture (solid)
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
  • For research use only

Catalog No. 50-004-49883 Supplier Medchemexpress LLC Supplier No. 250MGHYW005824
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,6-Dichloropurine Riboside (CAS 13276-52-3) is a chemical compound with a molecular weight of 321.12 and a purity of 98.97%. It appears as a white to off-white solid and is intended for research use only.

  • Chemical name: 9H-Purine, 2,6-dichloro-9-β-D-ribofuranosyl-
  • CAS number: 13276-52-3
  • Molecular formula: C10H10Cl2N4O4
  • Molecular weight: 321.12
  • Purity: 98.97%
  • Appearance: White to off-white solid
  • Storage conditions: -20°C, sealed storage, away from moisture (solid)
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.