Learn More
Medchemexpress LLC 9H-Purine-2,8-diamine, 9-[(3R)-1-(cyclopropylsulfonyl)-3-pyrrolidinyl]-N2-[4-(4-methyl-1-pipera | 2463200-44-2 | 99.3% | C29H35N9O2S | 10MG
SDP
Catalog No. 5000261233
Encompass_Preferred
EGFR-IN-11 is a fourth-generation EGFR tyrosine kinase inhibitor that selectively targets triple-mutant EGFR (L858R/T790M/C797S). It suppresses EGFR phosphorylation, induces apoptosis, and arrests cells in G0/G1; reported potency against the triple mutant is IC50 = 18 nM, and the compound is supplied for preclinical research.
  • High potency against EGFR L858R/T790M/C797S (IC50 18 nM).
  • Suppresses EGFR phosphorylation and induces apoptosis.
  • Causes cell cycle arrest at G0/G1 phase.
  • Reported activity across multiple cancer cell lines.
  • High chemical purity (99.3%).
Catalog No. 50-002-61233 Supplier Medchemexpress LLC Supplier No. HY13061610MG
EGFR-IN-11 is a fourth-generation EGFR tyrosine kinase inhibitor that selectively targets triple-mutant EGFR (L858R/T790M/C797S). It suppresses EGFR phosphorylation, induces apoptosis, and arrests cells in G0/G1; reported potency against the triple mutant is IC50 = 18 nM, and the compound is supplied for preclinical research.
  • High potency against EGFR L858R/T790M/C797S (IC50 18 nM).
  • Suppresses EGFR phosphorylation and induces apoptosis.
  • Causes cell cycle arrest at G0/G1 phase.
  • Reported activity across multiple cancer cell lines.
  • High chemical purity (99.3%).