Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9H-pyrido[3,4-b]indole-1-carboxaldehyde | 20127-63-3 | MFCD14706724 | 99.7% | 196.21 | C12H8N2O | 10 MG
SDP

Catalog No. 5000283425
Encompass_Preferred

1-Formyl-beta-carboline is a small-molecule beta-carboline alkaloid used as a research reagent. It inhibits Newcastle disease virus (NDV) infection by suppressing viral adsorption and entry, primarily through inhibition of the PI3K/Akt signaling pathway.

  • Reported antiviral activity against NDV via PI3K/Akt pathway inhibition.
  • High purity: 99.7% (HPLC), suitable for analytical and biological assays.
  • Chemical formula C12H8N2O and molecular weight 196.21.
  • CAS number 20127-63-3 for unambiguous identification.
  • Available in milligram-scale quantities for in vitro research use.

Catalog No. 50-002-83425 Supplier Medchemexpress LLC Supplier No. HYW13096510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Formyl-beta-carboline is a small-molecule beta-carboline alkaloid used as a research reagent. It inhibits Newcastle disease virus (NDV) infection by suppressing viral adsorption and entry, primarily through inhibition of the PI3K/Akt signaling pathway.

  • Reported antiviral activity against NDV via PI3K/Akt pathway inhibition.
  • High purity: 99.7% (HPLC), suitable for analytical and biological assays.
  • Chemical formula C12H8N2O and molecular weight 196.21.
  • CAS number 20127-63-3 for unambiguous identification.
  • Available in milligram-scale quantities for in vitro research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.