Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9H-pyrido[3,4-b]indole-1-carboxaldehyde | 20127-63-3 | MFCD14706724 | 99.7% | 196.21 g/mol | C12H8N2O | 25 MG
SDP

Catalog No. 5000283316
Encompass_Preferred

1-Formyl-β-carboline is a small-molecule alkaloid research compound reported to inhibit Akt and to exhibit antiviral activity. It is supplied as a solid for in vitro biochemical and cellular studies, with molecular formula C12H8N2O and molecular weight 196.21 g/mol.

  • CAS number: 20127-63-3.
  • Molecular formula: C12H8N2O; molecular weight: 196.21 g/mol.
  • Purity: 99.7% (reported by manufacturer).
  • Appearance: off-white to light brown solid.
  • Storage: sealed, away from moisture; solutions frozen for long-term storage.
  • Applications: biochemical assays and cellular studies targeting Akt-related pathways.

Catalog No. 50-002-83316 Supplier Medchemexpress LLC Supplier No. HYW13096525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Formyl-β-carboline is a small-molecule alkaloid research compound reported to inhibit Akt and to exhibit antiviral activity. It is supplied as a solid for in vitro biochemical and cellular studies, with molecular formula C12H8N2O and molecular weight 196.21 g/mol.

  • CAS number: 20127-63-3.
  • Molecular formula: C12H8N2O; molecular weight: 196.21 g/mol.
  • Purity: 99.7% (reported by manufacturer).
  • Appearance: off-white to light brown solid.
  • Storage: sealed, away from moisture; solutions frozen for long-term storage.
  • Applications: biochemical assays and cellular studies targeting Akt-related pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.