Learn More
Medchemexpress LLC 9H-Xanthen-9-one, 1,3,6,7-tetrahydroxy-2,8-bis(3-methyl-2-buten-1-yl)- | 31271-07-5 | 396.43 | 5 MG
SDP
Catalog No. 5000415912
Encompass_Preferred
Gamma-Mangostin is a novel competitive 5-hydroxytryptamine 2A (5-HT2A) receptor antagonist and a potent epoxidase 2 (COX-2) inhibitor, as well as a transthyroxin protein (TTR) profibrosis inhibitor. It binds to thyroxine (T4)-binding sites and stabilizes the TTR tetramer. This compound exhibits a range of biological activities and is used in the study of metabolic disorders.
  • Novel competitive 5-HT2A receptor antagonist
  • Potent COX-2 inhibitor
  • TTR profibrosis inhibitor
  • Stabilizes the TTR tetramer
  • Exhibits anti-inflammatory activity
  • Exhibits antibacterial activity
  • Exhibits antioxidant activity
  • Exhibits anticancer activities
  • Used in the study of metabolic disorders such as diabetes
Catalog No. 50-004-15912 Supplier Medchemexpress LLC Supplier No. HYN19575MG
Gamma-Mangostin is a novel competitive 5-hydroxytryptamine 2A (5-HT2A) receptor antagonist and a potent epoxidase 2 (COX-2) inhibitor, as well as a transthyroxin protein (TTR) profibrosis inhibitor. It binds to thyroxine (T4)-binding sites and stabilizes the TTR tetramer. This compound exhibits a range of biological activities and is used in the study of metabolic disorders.
  • Novel competitive 5-HT2A receptor antagonist
  • Potent COX-2 inhibitor
  • TTR profibrosis inhibitor
  • Stabilizes the TTR tetramer
  • Exhibits anti-inflammatory activity
  • Exhibits antibacterial activity
  • Exhibits antioxidant activity
  • Exhibits anticancer activities
  • Used in the study of metabolic disorders such as diabetes