Learn More
Cayman Chemical 9HODE cholesteryl ester 100ug
SDP
Catalog No. 502919143
Encompass_Preferred
A racemic monohydroxy fatty acid cholesteryl ester found in atherosclerotic lesions
Catalog No. 50-291-9143 Supplier Cayman Chemical Supplier No. 38401100 UG
A racemic monohydroxy fatty acid cholesteryl ester found in atherosclerotic lesions