Learn More
Ace Glass, Inc. 9in EXTENSION,MOD KILO STAND
SDP
Catalog No. 5000854733
Encompass
The Ace kilo-scale modular stand is a mobile platform for reactors and filtration vessels from 10 liters on up to 150 liters. Modular design allows a customer's existing stand to be modified to accept 200mm,300mm,400mm and 450mm flanged vessels as well as accommodating rod mounted or flange mounted stirring motors via interchangeable parts. Carriage lift is operated via a ratcheting handle or converted to an electric motor lift with the optional 12852-670 kit.
Catalog No. 50-008-54733 Supplier Ace Glass, Inc. Supplier No. 12852610
The Ace kilo-scale modular stand is a mobile platform for reactors and filtration vessels from 10 liters on up to 150 liters. Modular design allows a customer's existing stand to be modified to accept 200mm,300mm,400mm and 450mm flanged vessels as well as accommodating rod mounted or flange mounted stirring motors via interchangeable parts. Carriage lift is operated via a ratcheting handle or converted to an electric motor lift with the optional 12852-670 kit.