Learn More
Sklar Instruments Scissors, Merit Metzenbaum-Nelson Bariatric Scissors, Sklar 9 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094125 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Merit Metzenbaum-Nelson Bariatric Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA. Merit Metzenbaum-Nelson Bariatric Scissors are long dissecting scissors most commonly used in Bariatric Surgeries such as gastric bypass, gastric banding, and sleeve gastrectomy. They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue. Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors which make them ideal for use in thoracic procedures as well. These scissors are not recommended for cutting sutures or drains. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.
Catalog No. 50-109-4125 Supplier Sklar Instruments Supplier No. 97354
Scissors, Merit Metzenbaum-Nelson Bariatric Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Stainless Steel, EA. Merit Metzenbaum-Nelson Bariatric Scissors are long dissecting scissors most commonly used in Bariatric Surgeries such as gastric bypass, gastric banding, and sleeve gastrectomy. They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue. Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors which make them ideal for use in thoracic procedures as well. These scissors are not recommended for cutting sutures or drains. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.