Learn More
CELLTREAT Scientific Products 9 Inch Polypropylene Plasteur® Pasteur Pipet, Bulk Packed in Bags, Sterile pipette
SDP
Catalog No. 50828726
Encompass_Preferred
CELLTREAT Scientific Products 9"""" Polypropylen Plasteur Pipet. The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic. Fine tip is ideal for precision aspirating. Polypropylene provides the chemical resistivity needed for many applications. No more broken glass: more durable than standard glass pipets. Flexible plastic helps prevent glove or skin punctures. Will not shatter, eliminating losses due to broken glass. No damage during shipping and handling as compared to traditional glass pipets. Eliminate """"sharps"""" disposal. Can be autoclaved. Every case is printed with the lot number for quality traceability. Packaged sterile, 25 bulk packed/bag, 500/case.
Catalog No. 50-828-726 Supplier CELLTREAT Scientific Products Supplier No. 229286
CELLTREAT Scientific Products 9"""" Polypropylen Plasteur Pipet. The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic. Fine tip is ideal for precision aspirating. Polypropylene provides the chemical resistivity needed for many applications. No more broken glass: more durable than standard glass pipets. Flexible plastic helps prevent glove or skin punctures. Will not shatter, eliminating losses due to broken glass. No damage during shipping and handling as compared to traditional glass pipets. Eliminate """"sharps"""" disposal. Can be autoclaved. Every case is printed with the lot number for quality traceability. Packaged sterile, 25 bulk packed/bag, 500/case.