Learn More
Cayman Chemical 9-keto Fluprostenol isopropyl ester, Purity: ≥98%, CAS: 1219032-18-4, 10 mg
SDP
Catalog No. 502965785
Encompass_Preferred
An ester derivative of fluprostenol; a potential prodrug to 9-keto fluprostenol; a potential metabolite of fluprostenol isopropyl ester (travoprost)
Catalog No. 50-296-5785 Supplier Cayman Chemical Supplier No. 1678210 MG
An ester derivative of fluprostenol; a potential prodrug to 9-keto fluprostenol; a potential metabolite of fluprostenol isopropyl ester (travoprost)