Learn More
Gel Company 9LANE COMB,1.5MM,P/5,CBU09-150
Catalog No. NC1874343
Encompass_Preferred
9 lane comb, 1.5mm, for Mini-PROTEAN® 3 & Tetra Cell, pkg of 5 CBU09-150
Catalog No. NC1874343 Supplier Gel Company Supplier No. CBU09150
9 lane comb, 1.5mm, for Mini-PROTEAN® 3 & Tetra Cell, pkg of 5 CBU09-150