Learn More
S2 Media 9mL Buffered Peptone Water (BPW) - Poultry Rinse Fluid, 15mL Conical Tube, 50/box
Catalog No. NC3900332
Encompass_Preferred
Buffered Peptone Water (BPW) – Poultry Rinse Fluid is a non-selective pre-enrichment medium specifically formulated for the recovery and resuscitation of sublethal injured Salmonella spp. from poultry carcass rinse samples and other food matrices. It is used primarily in microbiological testing procedures prior to selective enrichment and isolation of target organisms. BPW provides a buffered environment and essential nutrients and allows for the repair of damaged cells, thereby improving the likelihood of detection in subsequent testing steps.• Product meets APHA, BAM, USDA, and WHO reference methods and performance criteria.• BPW should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. NC3900332 Supplier S2 Media Supplier No. 5515
Buffered Peptone Water (BPW) – Poultry Rinse Fluid is a non-selective pre-enrichment medium specifically formulated for the recovery and resuscitation of sublethal injured Salmonella spp. from poultry carcass rinse samples and other food matrices. It is used primarily in microbiological testing procedures prior to selective enrichment and isolation of target organisms. BPW provides a buffered environment and essential nutrients and allows for the repair of damaged cells, thereby improving the likelihood of detection in subsequent testing steps.• Product meets APHA, BAM, USDA, and WHO reference methods and performance criteria.• BPW should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.