Learn More
Analytical Sales & Services 9mm, 12 x 32, 1.5mL Clear Glass Wide Mouth Twist Vials for use with Robotic Arm Autosamplers
Catalog No. 5000806721
Encompass
Advantage™ 9mm, 12 x 32, 1.5mL Clear Glass Wide Mouth Twist Vials for use with Robotic Arm Autosamplers - Agilent, Waters, Leap, Perkin Elmer and Shimadzu
Catalog No. 50-008-06721 Supplier Analytical Sales & Services Supplier No. 11239
Advantage™ 9mm, 12 x 32, 1.5mL Clear Glass Wide Mouth Twist Vials for use with Robotic Arm Autosamplers - Agilent, Waters, Leap, Perkin Elmer and Shimadzu