Learn More
Analytical Sales & Services 9mm, 12 x 32, 1.7mL PP Wide Mouth Twist Vials
Catalog No. 5000805775
Encompass
Advantage™ 9mm, 12 x 32, 1.7mL Polypropylene Wide Mouth Twist Vials - for Robotic Arm Autosamplers: Agilent, Waters, Leap, Perkin Elmer and Shimadzu Good for Ion Chromatography
Catalog No. 50-008-05775 Supplier Analytical Sales & Services Supplier No. 88517CASE
Advantage™ 9mm, 12 x 32, 1.7mL Polypropylene Wide Mouth Twist Vials - for Robotic Arm Autosamplers: Agilent, Waters, Leap, Perkin Elmer and Shimadzu Good for Ion Chromatography