Learn More
Analytical Sales & Services 9mm, 12 x 32, 2mL Amber Glass Wide Mouth Twist Vials - Silanized
Catalog No. 5000805774
Encompass
Advantage™ 9mm, 12 x 32, 2mL Volume Amber Glass Wide Mouth Twist Vials - Silanized. For use with Robotic Arm Autosamplers: Agilent, Waters, Leap, Perkin Elmer and Shimadzu
Catalog No. 50-008-05774 Supplier Analytical Sales & Services Supplier No. 88119SCASE
Advantage™ 9mm, 12 x 32, 2mL Volume Amber Glass Wide Mouth Twist Vials - Silanized. For use with Robotic Arm Autosamplers: Agilent, Waters, Leap, Perkin Elmer and Shimadzu