Learn More
Analytical Sales & Services 9mm, 12 x 32, 2mL Amber Screw Vials with Marking Spot and Caps with PTFE/Silicone Liners
Catalog No. 5000805906
Encompass
Advantage™ Unassembled Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Screw Vials w/ White Marking Patch and 9mm Screw Caps with PTFE/Silicone Liners
Catalog No. 50-008-05906 Supplier Analytical Sales & Services Supplier No. 99006ACASE
Advantage™ Unassembled Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Amber Glass Screw Vials w/ White Marking Patch and 9mm Screw Caps with PTFE/Silicone Liners