Learn More
Analytical Sales & Services 9mm, 12 x 32, 2mL Clear Glass Wide Mouth Round Bottom Twist Vials
Catalog No. 5000807253
Encompass
Advantage™ 9mm, 12 x 32, 2mL Volume Clear Glass Wide Mouth Round Bottom Twist Vials - for use with Robotic Arm Autosamplers: Agilent, Waters, Leap, Perkin Elmer and Shimadzu
Catalog No. 50-008-07253 Supplier Analytical Sales & Services Supplier No. 88133CASE
Advantage™ 9mm, 12 x 32, 2mL Volume Clear Glass Wide Mouth Round Bottom Twist Vials - for use with Robotic Arm Autosamplers: Agilent, Waters, Leap, Perkin Elmer and Shimadzu