Learn More
Analytical Sales & Services 9mm, 12 x 32, 2mL Clear Screw Vials and Red Caps with PTFE/Silicone Liners
Catalog No. 5000805872
Encompass
Advantage™ Unassembled Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Clear Glass Screw Vials and 9mm Red Screw Caps with PTFE/Silicone Liners
Catalog No. 50-008-05872 Supplier Analytical Sales & Services Supplier No. 99006RCASE
Advantage™ Unassembled Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Clear Glass Screw Vials and 9mm Red Screw Caps with PTFE/Silicone Liners