Learn More
Analytical Sales & Services 9mm, 12 x 32, 2mL Clear Screw Vials and White Caps with PTFE/Silicone Liners
Catalog No. 5000805907
Encompass
Advantage™ Unassembled Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Clear Glass Screw Vials and 9mm White Screw Caps with PTFE/Silicone Liners
Catalog No. 50-008-05907 Supplier Analytical Sales & Services Supplier No. 99006WCASE
Add to Cart
Add to Cart
Advantage™ Unassembled Screw Vial Kit. Contains: 9mm, 12 x 32 2mL Clear Glass Screw Vials and 9mm White Screw Caps with PTFE/Silicone Liners