Learn More
Analytical Sales & Services 9mm, 12 x 32, Amber Glass Twist MicroVial with 300µL Glass Insert - Silanized
Catalog No. 5000806573
Encompass
Advantage™ 9mm, 12 x 32, Amber Glass Twist MicroVial with 300µL Volume Glass Insert - Silanized
Catalog No. 50-008-06573 Supplier Analytical Sales & Services Supplier No. 81233AS
Add to Cart
Add to Cart
Advantage™ 9mm, 12 x 32, Amber Glass Twist MicroVial with 300µL Volume Glass Insert - Silanized