Learn More
Analytical Sales & Services 9mm, 2mL, 12 x 32, Clear Glass Assembled Screw Vial Kit
Catalog No. 5000805557
Encompass
Advantage™ Pre-Assembled Vial Kit. Contains: 9mm, 12 x 32, 2mL Clear Glass Screw Vials and 9mm Screw Caps with Pre-Slit PTFE/Silicone Liners
Catalog No. 50-008-05557 Supplier Analytical Sales & Services Supplier No. 89006SLCASE
Advantage™ Pre-Assembled Vial Kit. Contains: 9mm, 12 x 32, 2mL Clear Glass Screw Vials and 9mm Screw Caps with Pre-Slit PTFE/Silicone Liners