Learn More
Analytical Sales & Services 9mm, 2mL Amber Wide Mouth Twist Vials and Blue Caps with Natural Rubber/PTFE Liners
Catalog No. 5000805091
Encompass
Advantage™ Pre-Assembled Vial Kit. Contains: 9mm, 2mL Amber Glass Wide Mouth Twist Vials and 9mm Blue Twist Caps with Natural Rubber/PTFE Liners
Catalog No. 50-008-05091 Supplier Analytical Sales & Services Supplier No. 58144CASE
Add to Cart
Add to Cart
Advantage™ Pre-Assembled Vial Kit. Contains: 9mm, 2mL Amber Glass Wide Mouth Twist Vials and 9mm Blue Twist Caps with Natural Rubber/PTFE Liners