Learn More
Analytical Sales & Services 9mm, 2mL Amber Wide Mouth Twist Vials and Blue Caps with PTFE/Silicone Liners
Catalog No. 5000804848
Encompass
Advantage™ Pre-Assembled Vial Kit. Contains: 9mm, 2mL Amber Glass Wide Mouth Twist Vials with Step and 9mm Blue Twist Caps with PTFE/Silicone Liners
Catalog No. 50-008-04848 Supplier Analytical Sales & Services Supplier No. 58128ACASE
Advantage™ Pre-Assembled Vial Kit. Contains: 9mm, 2mL Amber Glass Wide Mouth Twist Vials with Step and 9mm Blue Twist Caps with PTFE/Silicone Liners