Learn More
Analytical Sales & Services 9mm, 2mL Amber Wide Mouth Twist Vials with Marking Spot and Blue Caps with Natural Rubber/PTFE Liners
Catalog No. 5000804684
Encompass
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Amber Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Twist Caps with Natural Rubber/PTFE Liners
Catalog No. 50-008-04684 Supplier Analytical Sales & Services Supplier No. 58044ACASE
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Amber Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Twist Caps with Natural Rubber/PTFE Liners