Learn More
Analytical Sales & Services 9mm, 2mL Amber Wide Mouth Twist Vials with Marking Spot and Blue Caps with PTFE/Silcone/PTFE Liners
Catalog No. 5000806880
Encompass
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Amber Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Twist Caps with PTFE/Silcone/PTFE Liners
Catalog No. 50-008-06880 Supplier Analytical Sales & Services Supplier No. 58095ACASE
Advantage™ Unassembled Vial Kit. Contains: 9mm, 2mL Amber Glass Wide Mouth Twist Vials with Marking Spot and 9mm Blue Twist Caps with PTFE/Silcone/PTFE Liners