Learn More
Analytical Sales & Services 9mm, 2mL Amber Wide Mouth Twist Vials with Marking Spots and Blue Caps with PTFE/Silicone/PTFE Liners
Catalog No. 5000804531
Encompass
Advantage™ Pre-Assembled Vial Kit\n\n• Volume: 2mL\n• Vial Opening: 9mm OD\n• Overall Dimensions: 12 x 32mm\n• Style: Wide Mouth Twist Vials with Marking Spots\n• Color: Amber\n• Includes: Blue Twist Caps with PTFE/Silicone/PTFE Liners\n\nQty/Pkg: 1000 (case, 10 packs of 100)
Catalog No. 50-008-04531 Supplier Analytical Sales & Services Supplier No. 58125ACASE
Advantage™ Pre-Assembled Vial Kit\n\n• Volume: 2mL\n• Vial Opening: 9mm OD\n• Overall Dimensions: 12 x 32mm\n• Style: Wide Mouth Twist Vials with Marking Spots\n• Color: Amber\n• Includes: Blue Twist Caps with PTFE/Silicone/PTFE Liners\n\nQty/Pkg: 1000 (case, 10 packs of 100)